Ethylene glycol, 95%, pure, Thermo Scientific Chemicals
Ethylene glycol, 95%, pure, Thermo Scientific Chemicals
Ethylene glycol, 95%, pure, Thermo Scientific Chemicals
Ethylene glycol, 95%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylene glycol, 95%, pure, Thermo Scientific Chemicals
CAS: 107-21-1 | C2H6O2 | 62.068 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
1 kg
Packaging:
Glass Bottle
Catalog number FSA410010025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 kg
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS77-79-2
IUPAC Name2,5-dihydro-1λ⁶-thiophene-1,1-dione
Molecular FormulaC4H6O2S
InChI KeyMBDNRNMVTZADMQ-UHFFFAOYSA-N
SMILESO=S1(=O)CC=CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Adhering powder or crystals and/or chunks
Appearance (Color)White to off-white
Infrared spectrumConforms
GC>=97.5 %
Melting point63°C to 67°C