Ethylene dimethacrylate, 98%, stabilized, Thermo Scientific Chemicals
Ethylene dimethacrylate, 98%, stabilized, Thermo Scientific Chemicals
Ethylene dimethacrylate, 98%, stabilized, Thermo Scientific Chemicals
Ethylene dimethacrylate, 98%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylene dimethacrylate, 98%, stabilized, Thermo Scientific Chemicals
CAS: 97-90-5 | C10H14O4 | 198.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA409922500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS615-59-8
IUPAC Name1,4-dibromo-2-methylbenzene
Molecular FormulaC7H6Br2
InChI KeyQKEZTJYRBHOKHH-UHFFFAOYSA-N
SMILESCC1=CC(Br)=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Refractive Index1.6005-1.6045 @ 20?C