Ethylenebis(oxyethylenenitrilo)tetraacetic acid, 98%, Thermo Scientific Chemicals
Ethylenebis(oxyethylenenitrilo)tetraacetic acid, 98%, Thermo Scientific Chemicals
Ethylenebis(oxyethylenenitrilo)tetraacetic acid, 98%, Thermo Scientific Chemicals
Ethylenebis(oxyethylenenitrilo)tetraacetic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethylenebis(oxyethylenenitrilo)tetraacetic acid, 98%, Thermo Scientific Chemicals
CAS: 67-42-5 | C14H24N2O10 | 380.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number FSA409911000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS938-18-1
IUPAC Name2,4,6-trimethylbenzoyl chloride
Molecular FormulaC10H11ClO
InChI KeyUKRQMDIFLKHCRO-UHFFFAOYSA-N
SMILESCC1=CC(C)=C(C(Cl)=O)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless to yellow
Refractive Index1.5270-1.5310 @ 20?C
Assay (Titration ex Chloride)>98.0 to <102.0% (liberated Cl⁻ from reaction with Pyridine)
FormLiquid
Assay (GC)>98.0%
Chelation/Complexation reagent
RUO – Research Use Only