1,4-Dioxane, 99+%, ACS reagent, stabilized, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
1,4-Dioxane, 99+%, ACS reagent, stabilized, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
1,4-Dioxane, 99+%, ACS reagent, stabilized, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
1,4-Dioxane, 99+%, ACS reagent, stabilized, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Dioxane, 99+%, ACS reagent, stabilized, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals
CAS: 123-91-1 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Packaging:
View all
EcoSafPak™ Glass Bottle
Catalog number FSA408825000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
EcoSafPak™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS29558-78-9
IUPAC Name4'-iodo-[1,1'-biphenyl]-4-ol
Molecular FormulaC12H9IO
InChI KeyWJXIAMCDWSUSEI-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Melting Point196-203°C