2,4-Dimethylphenol, 99%, Thermo Scientific Chemicals
2,4-Dimethylphenol, 99%, Thermo Scientific Chemicals
2,4-Dimethylphenol, 99%, Thermo Scientific Chemicals
2,4-Dimethylphenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,4-Dimethylphenol, 99%, Thermo Scientific Chemicals
CAS: 105-67-9 | C8H10O | 122.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA408451000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 4-Dimethylphenol
Melting Point21.0°C to 23.0°C
CAS105-67-9
CAS Min %98.5
ColorColorless to Yellow