Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
Thermo Scientific Chemicals
Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution
CAS: 140-01-2 | C14H18N3Na5O10 | 503.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
3 kg
Packaging:
Glass bottle
Catalog number FSA407290010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3647-69-6
IUPAC Namehydrogen 4-(2-chloroethyl)morpholine chloride
Molecular FormulaC6H13Cl2NO
InChI KeyNBJHDLKSWUDGJG-UHFFFAOYSA-N
SMILES[H+].[Cl-].ClCCN1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream
Assay (Titration ex Chloride)≥98.5 to ≤101.5%
Solution TestClear (5% in water)
Chelation/Complexation reagent
RUO – Research Use Only