Copper(I) chloride, 90+%, ACS reagent, Thermo Scientific Chemicals
Copper(I) chloride, 90+%, ACS reagent, Thermo Scientific Chemicals
Copper(I) chloride, 90+%, ACS reagent, Thermo Scientific Chemicals
Copper(I) chloride, 90+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Copper(I) chloride, 90+%, ACS reagent, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA405871000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS629-25-4
IUPAC Namesodium dodecanoate
Molecular FormulaC12H23NaO2
InChI KeyBTURAGWYSMTVOW-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Water=<3 %
Appearance (Color)White to light yellow