Copper(II) oxide, ACS reagent, powder, Thermo Scientific Chemicals
Copper(II) oxide, ACS reagent, powder, Thermo Scientific Chemicals
Copper(II) oxide, ACS reagent, powder, Thermo Scientific Chemicals
Copper(II) oxide, ACS reagent, powder, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Copper(II) oxide, ACS reagent, powder, Thermo Scientific Chemicals
CAS: 1317-38-0 | CuO | 79.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
Packaging:
View all
Plastic bottle
Catalog number FSA405862500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS57-50-1
IUPAC Name(2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC12H22O11
InChI KeyCZMRCDWAGMRECN-PWPRYFECNA-N
SMILESOC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
DNaseNone detected
RNaseNone detected