o-Cresol, 99%, AcroSeal™, Thermo Scientific Chemicals
o-Cresol, 99%, AcroSeal™, Thermo Scientific Chemicals
o-Cresol, 99%, AcroSeal™, Thermo Scientific Chemicals
o-Cresol, 99%, AcroSeal™, Thermo Scientific Chemicals
o-Cresol, 99%, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
o-Cresol, 99%, AcroSeal™, Thermo Scientific Chemicals
CAS: 95-48-7 | C7H8O | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA405731000
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS137-08-6
IUPAC Namecalcium bis(3-[(2R)-2,4-dihydroxy-3,3-dimethylbutanamido]propanoate)
Molecular FormulaC18H32CaN2O10
InChI KeyFAPWYRCQGJNNSJ-DXHDTSSINA-L
SMILES[Ca++].CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O.CC(C)(CO)[C@@H](O)C(=O)NCCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation+25.0° to +27.5° (20°C, 589 nm) (c=5, H2O)
Specific optical rotationOn dried substance
Loss on drying=<5.0 % (105°C, 3 h)
Heavy metals (as Pb)=<20 ppm
Appearance (Color)White to almost white