Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals
CAS: 7440-44-0 | C | 12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number FSA404035000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3599-32-4
IUPAC Namesodium 2-[(5E)-7-[(2E)-1,1-dimethyl-3-(4-sulfonatobutyl)-1H,2H,3H-benzo[e]indol-2-ylidene]hepta-1,3,5-trien-1-yl]-1,1-dimethyl-3-(4-sulfonatobutyl)-1H-benzo[e]indol-3-ium
Molecular FormulaC43H47N2NaO6S2
InChI KeyMOFVSTNWEDAEEK-UHFFFAOYSA-M
SMILES[Na+].CC1(C)C(=C/C=C/C=CC=CC2=[N+](CCCCS([O-])(=O)=O)C3=CC=C4C=CC=CC4=C3C2(C)C)N(CCCCS([O-])(=O)=O)C2=CC=C3C=CC=CC3=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
UV Lambda max784 nm (in Methanol)
Molar absorption coefficient>=230000 l/(mol.cm)
Appearance (Color)Green to brown
Appearance (Form)Crystalline powder or crystals
Filter aid
RUO – Research Use Only