CAB-O-SIL™ M-5, Scintillation Grade
CAB-O-SIL™ M-5, Scintillation Grade
CAB-O-SIL™ M-5, Scintillation Grade
CAB-O-SIL™ M-5, Scintillation Grade
Thermo Scientific Chemicals
CAB-O-SIL™ M-5, Scintillation Grade
CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA403731500150 gPlastic Bottle
Catalog number FSA403731500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
150 g
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-47-3
IUPAC Namechromium
Molecular FormulaCr
InChI KeyVYZAMTAEIAYCRO-UHFFFAOYSA-N
SMILES[Cr]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPieces, 3-12mm
Total Metal Impurities≤0.8%
For column chromatography, For fluorescence/luminescence spectroscopy
RUO – Research Use Only