Bis(2-hydroxyethyl)dimethylammonium chloride, 99%
Bis(2-hydroxyethyl)dimethylammonium chloride, 99%
Bis(2-hydroxyethyl)dimethylammonium chloride, 99%
Bis(2-hydroxyethyl)dimethylammonium chloride, 99%
Thermo Scientific Chemicals
Bis(2-hydroxyethyl)dimethylammonium chloride, 99%
CAS: 38402-02-7 | C6H16ClNO2 | 169.65 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA40253025025 gGlass bottle
Catalog number FSA402530250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialBis(2-hydroxyethyl)dimethylammonium chloride
Name Note99%
CAS38402-02-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
View more
Phase transfer catalyst
RUO – Research Use Only