Bis(2-ethylhexyl) hydrogen phosphate, 95%, Thermo Scientific Chemicals
Bis(2-ethylhexyl) hydrogen phosphate, 95%, Thermo Scientific Chemicals
Bis(2-ethylhexyl) hydrogen phosphate, 95%, Thermo Scientific Chemicals
Bis(2-ethylhexyl) hydrogen phosphate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bis(2-ethylhexyl) hydrogen phosphate, 95%, Thermo Scientific Chemicals
CAS: 298-07-7 | C16H35O4P | 322.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Glass Bottle
Catalog number FSA402470010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS877-24-7
IUPAC Namepotassium 2-carboxybenzoate
Molecular FormulaC8H5KO4
InChI KeyIWZKICVEHNUQTL-UHFFFAOYSA-M
SMILES[K+].OC(=O)C1=CC=CC=C1C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay99.95 to 100.05 %
pH3.8 to 4 (5 % at 20°C)
Chlorine compounds=<30 ppm (as Cl)
Iron (Fe)=<5 ppm
Sulfate (SO4)=<60 ppm