Azelaic acid, 98%, Thermo Scientific Chemicals
Azelaic acid, 98%, Thermo Scientific Chemicals
Azelaic acid, 98%, Thermo Scientific Chemicals
Azelaic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Azelaic acid, 98%, Thermo Scientific Chemicals
CAS: 123-99-9 | C9H16O4 | 188.223 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
View all
Glass bottle
Catalog number FSA401521000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS140690-56-8
IUPAC Name1-(bromomethyl)-2,4-bis(trifluoromethyl)benzene
Molecular FormulaC9H5BrF6
InChI KeySWFFFUJOWAJJCH-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=C(CBr)C(=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.4480-1.4530 @ 20?C
Assay (GC)≥96.0%