Aluminon, ACS reagent, Thermo Scientific Chemicals
Aluminon, ACS reagent, Thermo Scientific Chemicals
Aluminon, ACS reagent, Thermo Scientific Chemicals
Aluminon, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Aluminon, ACS reagent, Thermo Scientific Chemicals
CAS: 569-58-4 | C22H23N3O9 | 473.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA401501000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire, 1.5mm (0.06in) diameter.
Appearance (Color)Silver
Assay from Supplier's CofA≥99.998% (metals basis)