2-Amino-5-nitrophenol, 95%, Thermo Scientific Chemicals
2-Amino-5-nitrophenol, 95%, Thermo Scientific Chemicals
2-Amino-5-nitrophenol, 95%, Thermo Scientific Chemicals
2-Amino-5-nitrophenol, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Amino-5-nitrophenol, 95%, Thermo Scientific Chemicals
CAS: 121-88-0 | C6H6N2O3 | 154.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number FSA400900500
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS88-17-5
IUPAC Name2-(trifluoromethyl)aniline
Molecular FormulaC7H6F3N
InChI KeyVBLXCTYLWZJBKA-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=98.5 %
Infrared spectrumConforms
Refractive index1.4800 to 1.4830 (20°C, 589 nm)