tert-Butanol, Thermo Scientific Chemicals
tert-Butanol, Thermo Scientific Chemicals
tert-Butanol, Thermo Scientific Chemicals
tert-Butanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
tert-Butanol, Thermo Scientific Chemicals
CAS: 75-65-0 | C4H10O | 74.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Plastic Bottle
Catalog number FSA390690010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS135610-90-1
IUPAC Name2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-[1-(triphenylmethyl)-1H-imidazol-4-yl]propanoic acid
Molecular FormulaC40H33N3O4
InChI KeyXXMYDXUIZKNHDT-UHFFFAOYNA-N
SMILESOC(=O)C(CC1=CN(C=N1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%