1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, ACS reagent, Thermo Scientific Chemicals
CAS: 872-50-4 | C5H9NO | 99.133 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number FSA390682500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS14104-20-2
IUPAC Namesilver(1+) tetrafluoroboranuide
Molecular FormulaAgBF4
InChI KeyCCAVYRRHZLAMDJ-UHFFFAOYSA-N
SMILES[Ag+].F[B-](F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to very pale cream
Assay (Argentometric Titration)≥98.5 to ≤101.5%
Suitable for battery materials development.
RUO – Research Use Only