Ammonium hydroxide, for analysis, 20% solution in water, Thermo Scientific Chemicals
Ammonium hydroxide, for analysis, 20% solution in water, Thermo Scientific Chemicals
Ammonium hydroxide, for analysis, 20% solution in water, Thermo Scientific Chemicals
Ammonium hydroxide, for analysis, 20% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium hydroxide, for analysis, 20% solution in water, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Glass Bottle
Catalog number FSA390070025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS13446-34-9
IUPAC Namemanganese(2+) tetrahydrate dichloride
Molecular FormulaCl2H8MnO4
InChI KeyCNFDGXZLMLFIJV-UHFFFAOYSA-L
SMILESO.O.O.O.[Cl-].[Cl-].[Mn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble matter=<0.005 %
Potassium (K)=<0.01 %
Appearance (Form)Crystals
pH3.5 to 6.0 (5% soln. at 25°C)
Magnesium (Mg)=<0.005 %