Pumice stone, granular (approx. 1-5mm), Thermo Scientific Chemicals
Pumice stone, granular (approx. 1-5mm), Thermo Scientific Chemicals
Pumice stone, granular (approx. 1-5mm), Thermo Scientific Chemicals
Pumice stone, granular (approx. 1-5mm), Thermo Scientific Chemicals
Alfa Aesar
Pumice stone, granular (approx. 1-5mm), Thermo Scientific Chemicals
CAS: 1332-09-8 | C28H37FO7 | 509.626 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic Bottle
Catalog number FSA390040010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS164513-38-6
IUPAC Name5-bromo-4-methyl-1,2-dihydropyridin-2-one
Molecular FormulaC6H6BrNO
InChI KeyBYGNXKDNIFHZHQ-UHFFFAOYSA-N
SMILESCC1=CC(=O)NC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%