Ammonium hydroxide, for analysis, 25% free ammonia, solution in water, Thermo Scientific Chemicals
Ammonium hydroxide, for analysis, 25% free ammonia, solution in water, Thermo Scientific Chemicals
Ammonium hydroxide, for analysis, 25% free ammonia, solution in water, Thermo Scientific Chemicals
Ammonium hydroxide, for analysis, 25% free ammonia, solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium hydroxide, for analysis, 25% free ammonia, solution in water, Thermo Scientific Chemicals
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA390030010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS54723-94-3
SpecificationsSpecification SheetSpecification Sheet
Impurity contentCa < 0.02%
Impurity contentSO₄ < 0.025%
Insoluble material< 0.01%
Trace Metals(as Pb) < 0.005%
Impurity contentCl < 0.05%