FormCrystals or powder or crystalline powder or flakes
Appearance (Color)White to pale cream
Assay (GC)≥98.5% (non-U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Water Content (Karl Fischer Titration)≤1% (non-U.S. specification)
View more