n-Pentane, for analysis, Thermo Scientific Chemicals
n-Pentane, for analysis, Thermo Scientific Chemicals
n-Pentane, for analysis, Thermo Scientific Chemicals
n-Pentane, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Pentane, for analysis, Thermo Scientific Chemicals
CAS: 109-66-0 | C5H12 | 72.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number FSA383900010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS1083-30-3
IUPAC Name1,3-diphenylpropan-1-one
Molecular FormulaC15H14O
InChI KeyQGGZBXOADPVUPN-UHFFFAOYSA-N
SMILESO=C(CCC1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown
FormCrystalline powder
Assay (GC)>97.5%
Clarity(5% w/v solution in acetone) : Transparent to almost transparent by JIS K 8001 5.2
For analysis
RUO – Research Use Only