Check Order Status
3-methylthiophene-2-carbonyl Chloride, 98%, Thermo Scientific™
Element Search
Structure Search
Thermo Scientific Chemicals
3-methylthiophene-2-carbonyl Chloride, 98%, Thermo Scientific™
Have Questions?
Change view
Quantity:
10 g
Packaging:
Glass bottle
Catalog number
FSA380400100
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material
3-Methylthiophene-2-carbonyl chloride
CAS
61341-26-2
CAS Min %
97.5
Health Hazard 1
GHS Signal Word:
Danger
CAS Max %
100.0