Tetraethylammonium perchlorate, 0.2M aqueous solution, Thermo Scientific Chemicals
Tetraethylammonium perchlorate, 0.2M aqueous solution, Thermo Scientific Chemicals
Tetraethylammonium perchlorate, 0.2M aqueous solution, Thermo Scientific Chemicals
Tetraethylammonium perchlorate, 0.2M aqueous solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetraethylammonium perchlorate, 0.2M aqueous solution, Thermo Scientific Chemicals
CAS: 2567-83-1 | C8H20ClNO4 | 229.70 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Plastic bottle
Catalog number FSA379801000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS73852-19-4
IUPAC Name[3,5-bis(trifluoromethyl)phenyl]boronic acid
Molecular FormulaC8H5BF6O2
InChI KeyBPTABBGLHGBJQR-UHFFFAOYSA-N
SMILESOB(O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.0%
Proton NMRConforms to structure
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream
Assay (HPLC)≥97.0%
Oxidation reagent, Phase transfer catalyst
RUO – Research Use Only