DL-Lactide, 99%, Thermo Scientific Chemicals
DL-Lactide, 99%, Thermo Scientific Chemicals
DL-Lactide, 99%, Thermo Scientific Chemicals
DL-Lactide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Lactide, 99%, Thermo Scientific Chemicals
CAS: 95-96-5 | C6H8O4 | 144.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA376851000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS78-98-8
IUPAC Name2-oxopropanal
Molecular FormulaC3H4O2
InChI KeyAIJULSRZWUXGPQ-UHFFFAOYSA-N
SMILESCC(=O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Refractive index1.3910 to 1.4200 (20°C, 589 nm)
Appearance (Color)Clear yellow to brown
Titration with NaOH30 to 45 %