Nordihydroguaiaretic acid, 95%, Thermo Scientific Chemicals
Nordihydroguaiaretic acid, 95%, Thermo Scientific Chemicals
Nordihydroguaiaretic acid, 95%, Thermo Scientific Chemicals
Nordihydroguaiaretic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nordihydroguaiaretic acid, 95%, Thermo Scientific Chemicals
CAS: 500-38-9 | C18H22O4 | 302.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
Packaging:
Glass bottle
Catalog number FSA376305000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS98-79-3
IUPAC Name5-oxopyrrolidine-2-carboxylic acid
Molecular FormulaC5H7NO3
InChI KeyODHCTXKNWHHXJC-UHFFFAOYNA-N
SMILESOC(=O)C1CCC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with NaOH>=97.5 % (On dry substance)
Specific optical rotation-26° to -29° (20°C, 589 nm) (c=10, 1N NaOH)
Appearance (Form)Crystalline powder
Specific optical rotationon dry substance