Silica gel, C18-RP, 23%C, ca.1.2mmol/g, part. size 40-63μm
Silica gel, C18-RP, 23%C, ca.1.2mmol/g, part. size 40-63μm
Thermo Scientific Chemicals
Silica gel, C18-RP, 23%C, ca.1.2mmol/g, part. size 40-63μm
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Catalog number FSA360110500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS18156-74-6
IUPAC Name1-(trimethylsilyl)-1H-imidazole
Molecular FormulaC6H12N2Si
InChI KeyYKFRUJSEPGHZFJ-UHFFFAOYSA-N
SMILESC[Si](C)(C)N1C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.4740-1.4790 @ 20?C
FormLiquid
For column chromatography
RUO – Research Use Only