Nocodazole, 98%, Thermo Scientific Chemicals
Nocodazole, 98%, Thermo Scientific Chemicals
Nocodazole, 98%, Thermo Scientific Chemicals
Nocodazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nocodazole, 98%, Thermo Scientific Chemicals
CAS: 31430-18-9 | C14H11N3O3S | 301.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
10 mg
Packaging:
Glass bottle
Catalog number FSA358240500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS141-28-6
IUPAC Name1,6-diethyl hexanedioate
Molecular FormulaC10H18O4
InChI KeyVIZORQUEIQEFRT-UHFFFAOYSA-N
SMILESCCOC(=O)CCCCC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
Refractive Index1.4265-1.4295 @ 20?C
Assay (GC)≥98.5%