1-Ethyl-3-methylimidazolium chloride, 97%, Thermo Scientific Chemicals
1-Ethyl-3-methylimidazolium chloride, 97%, Thermo Scientific Chemicals
1-Ethyl-3-methylimidazolium chloride, 97%, Thermo Scientific Chemicals
1-Ethyl-3-methylimidazolium chloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Ethyl-3-methylimidazolium chloride, 97%, Thermo Scientific Chemicals
CAS: 65039-09-0 | C6H11ClN2 | 146.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA354081000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS638-53-9
IUPAC Nametridecanoic acid
Molecular FormulaC13H26O2
InChI KeySZHOJFHSIKHZHA-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=96.0 %
Melting point40°C to 45°C
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder or flakes and/or chunks
Infrared spectrumConforms
Ionic liquid
RUO – Research Use Only