Acetone-d6, for NMR, with 0.03% TMS, 99.9% atom D, Thermo Scientific Chemicals
Acetone-d6, for NMR, with 0.03% TMS, 99.9% atom D, Thermo Scientific Chemicals
Acetone-d6, for NMR, with 0.03% TMS, 99.9% atom D, Thermo Scientific Chemicals
Acetone-d6, for NMR, with 0.03% TMS, 99.9% atom D, Thermo Scientific Chemicals
Acetone-d6, for NMR, with 0.03% TMS, 99.9% atom D, Thermo Scientific Chemicals
CAS: 666-52-4 | C3H6O | 64.117 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
10 mL
Packaging:
Glass Bottle
Catalog number FSA351380500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS856405-77-1
IUPAC Namedi-tert-butyl[5-(di-tert-butylphosphanyl)-9,9-dimethyl-9H-xanthen-4-yl]phosphane
Molecular FormulaC31H48OP2
InChI KeyZEIZANJFJXHMNS-UHFFFAOYSA-N
SMILESCC(C)(C)P(C1=CC=CC2=C1OC1=C(C=CC=C1P(C(C)(C)C)C(C)(C)C)C2(C)C)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay by NMR>98.5% by 31P NMR
Proton NMRConforms to structure
FormPowder
for NMR
RUO – Research Use Only