Perchloric acid, pure, 0.1M solution in acetic acid, Thermo Scientific Chemicals
Perchloric acid, pure, 0.1M solution in acetic acid, Thermo Scientific Chemicals
Perchloric acid, pure, 0.1M solution in acetic acid, Thermo Scientific Chemicals
Perchloric acid, pure, 0.1M solution in acetic acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Perchloric acid, pure, 0.1M solution in acetic acid, Thermo Scientific Chemicals
CAS: 7601-90-3 | ClHO4 | 100.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Plastic Bottle
Catalog number FSA348630010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS722-92-9
IUPAC Name2-(4-aminophenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol
Molecular FormulaC9H7F6NO
InChI KeyTZEJXCIGVMTMDY-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1)C(O)(C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
Assay (GC)≥95.0%
FormPowder
Melting Point (clear melt)145.0-154.0?C
Oxidation reagent
RUO – Research Use Only