N,N-Dimethylformamide, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.8%, C3H7NO, CAS Number-68-12-2, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dwumetyloformamid, dimethylformamide, dmfa, dimethyl formamide, dimetilformamide, dmf, dimethylformamid, 1L, 0.01 % max., Vapor Pressure: 4.9mbar at 20 deg.C, 04, 58
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
500 mL
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA348430010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS603-76-9
IUPAC Name1-methyl-1H-indole
Molecular FormulaC9H9N
InChI KeyBLRHMMGNCXNXJL-UHFFFAOYSA-N
SMILESCN1C=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.6050 to 1.6080 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.0 %