Tetraacetylethylenediamine, 90%, Thermo Scientific Chemicals
Tetraacetylethylenediamine, 90%, Thermo Scientific Chemicals
Tetraacetylethylenediamine, 90%, Thermo Scientific Chemicals
Tetraacetylethylenediamine, 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetraacetylethylenediamine, 90%, Thermo Scientific Chemicals
CAS: 10543-57-4 | C10H16N2O4 | 228.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Plastic bottle
Catalog number FSA344592500
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS110-86-1
IUPAC Namepyridine
Molecular FormulaC5H5N
InChI KeyJUJWROOIHBZHMG-UHFFFAOYSA-N
SMILESC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.1 % (Coulometric)
Residue after evaporation=<0.02 %
Refractive index1.5080 to 1.5110 (20°C, 589 nm)
Appearance (Form)Clear liquid
GC>=99.0 %