Carbamazepine, 98%
Carbamazepine, 98%
Carbamazepine, 98%
Thermo Scientific Chemicals
Carbamazepine, 98%
Carbamazepine (Carbamazepen), CAS # 298-46-4, is an aromatic ring of the dibenzoazepine class that acts as a sodium channel blocker.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
25 g
Packaging:
Catalog number FSA344360050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS27176-10-9
IUPAC Nametripotassium hydrate phosphate
Molecular FormulaH2K3O5P
InChI KeyRMNIZOOYFMNEJJ-UHFFFAOYSA-K
SMILESO.[K+].[K+].[K+].[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals or granules
Loss on ignition=<13 % (800°C)
Iron (Fe)=<20 ppm
Appearance (Color)White
Titration excess HCl>=95 %
View more
General Description
  • Carbamazepine is an aromatic ring of the dibenzoazepine class that acts as a sodium channel blocker
  • It is a tricyclic antidepressant-like drug that reduces post-tetanic potentiation
Applications
  • Carbamazepine can be used as a sodium channel blocker and as a glutamate-transporter agonist to reduce neural activity and produce inhibitory effects in the brain
  • Carbamazepine is useful in the research of pain, epilepsy, and manic depression
RUO – Research Use Only
General References:
  1. Ben-Menachem, E.; Grebe, H.P.; Terada, K.; Jensen, L.; Li, T.; De Backer, M.; Steiniger-Brach, B.; Gasalla, T.; Brock, M.; Biton, V. Long-term safety and efficacy of lacosamide and controlled-release carbamazepine monotherapy in patients with newly diagnosed epilepsy. Epilepsia. 2019, 60(12), 2437-2447. Erratum in: Epilepsia. 2021, 62(4), 1039.
  2. Schmitz, B.; Dimova, S.; Zhang, Y.; Chellun, D.; De Backer, M.; Gasalla, T. Tolerability and efficacy of lacosamide and controlled-release carbamazepine monotherapy in patients with newly diagnosed epilepsy and concomitant psychiatric conditions: Post hoc analysis of a prospective, randomized, double-blind trial. Epilepsy Res. 2020, 159, 106220.