Polysorbate 40, Thermo Scientific Chemicals
Polysorbate 40, Thermo Scientific Chemicals
Polysorbate 40, Thermo Scientific Chemicals
Polysorbate 40, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Polysorbate 40, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA334142500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1304-76-3
IUPAC Namedibismuth(3+) trioxidandiide
Molecular FormulaBi2O3
InChI KeyTYIXMATWDRGMPF-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Bi+3].[Bi+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Surface Area12-24 m²/g
Assay (unspecified)99.5% min