2-Decanol, 98%, Thermo Scientific Chemicals
2-Decanol, 98%, Thermo Scientific Chemicals
2-Decanol, 98%, Thermo Scientific Chemicals
2-Decanol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Decanol, 98%, Thermo Scientific Chemicals
CAS: 1120-06-5 | C10H22O | 158.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number FSA331600050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS499-81-0
IUPAC Namepyridine-3,5-dicarboxylic acid
Molecular FormulaC7H5NO4
InChI KeyMPFLRYZEEAQMLQ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(=CN=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder or crystals or crystalline powder
Assay (HPLC)≥97.5%