(±)-Verapamil hydrochloride, 99+%
(±)-Verapamil hydrochloride, 99+%
(±)-Verapamil hydrochloride, 99+%
(±)-Verapamil hydrochloride, 99+%
Thermo Scientific Chemicals
(±)-Verapamil hydrochloride, 99+%
CAS: 152-11-4 | C27H38N2O4·HCl | 491.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA329330050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS24720-64-7
IUPAC Name2-(triphenyl-λ⁵-phosphanylidene)propanal
Molecular FormulaC21H19OP
InChI KeyVHUQEFAWBCDBSC-UHFFFAOYSA-N
SMILESCC(C=O)=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Appearance (Form)Powder
Infrared spectrumConforms
NMR (31P-NMR)>=97.5 %
NMR (1H-NMR)>=97.5 %