1,2-Diiodotetrafluorobenzene, 99%, Thermo Scientific Chemicals
1,2-Diiodotetrafluorobenzene, 99%, Thermo Scientific Chemicals
1,2-Diiodotetrafluorobenzene, 99%, Thermo Scientific Chemicals
1,2-Diiodotetrafluorobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,2-Diiodotetrafluorobenzene, 99%, Thermo Scientific Chemicals
CAS: 2708-97-6 | C6F4I2 | 401.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number FSA311690050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS99-10-5
IUPAC Name3,5-dihydroxybenzoic acid
Molecular FormulaC7H6O4
InChI KeyUYEMGAFJOZZIFP-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(O)=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to cream or pale pink
Identification (FTIR)Conforms
Assay (HPLC)≥96.0%
Melting Point (clear melt)235-242?C