Isopropyl-beta-D-thiogalactopyranoside, 99%, for biochemistry, dioxane-free
Isopropyl-beta-D-thiogalactopyranoside, 99%, for biochemistry, dioxane-free
Isopropyl-beta-D-thiogalactopyranoside, 99%, for biochemistry, dioxane-free
Isopropyl-beta-D-thiogalactopyranoside, 99%, for biochemistry, dioxane-free
Isopropyl-beta-D-thiogalactopyranoside, 99%, for biochemistry, dioxane-free
CAS: 367-93-1 | C9H18O5S | 238.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
10 g
Packaging:
Glass bottle
Catalog number FSA302790250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS106-54-7
IUPAC Name4-chlorobenzene-1-thiol
Molecular FormulaC6H5ClS
InChI KeyVZXOZSQDJJNBRC-UHFFFAOYSA-N
SMILESSC1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or lumps or fused solid
Melting Point (clear melt)48.0-57.0°C
Appearance (Color)White to pale cream
Assay (GC)≥96.0%