Maltitol, 95%
Maltitol, 95%
Maltitol, 95%
Thermo Scientific Chemicals
Maltitol, 95%
CAS: 585-88-6 | C12H24O11 | 344.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
1 kg
Packaging:
Glass bottle
Catalog number FSA295802500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS62-44-2
IUPAC NameN-(4-ethoxyphenyl)acetamide
Molecular FormulaC10H13NO2
InChI KeyCPJSUEIXXCENMM-UHFFFAOYSA-N
SMILESCCOC1=CC=C(NC(C)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder or flakes
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Melting Point (clear melt)131.0-140.0?C