2,2,4-Trimethylpentane, 99+%, pure, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, 99+%, pure, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, 99+%, pure, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, 99+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2,4-Trimethylpentane, 99+%, pure, Thermo Scientific Chemicals
CAS: 540-84-1 | C8H18 | 114.232 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Glass Bottle
Catalog number FSA295720025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS7732-18-5
IUPAC Namewater
Molecular FormulaH2O
InChI KeyXLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILESO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Conductivity=<1 µS/cm
Residue after evaporation=<0.0003 %
UVat 200 nm A: =<0.05
UVat 210 nm A: =<0.009