Glycerol, for analysis, 86-88% wt% aqueous solution, Thermo Scientific Chemicals
Glycerol, for analysis, 86-88% wt% aqueous solution, Thermo Scientific Chemicals
Glycerol, for analysis, 86-88% wt% aqueous solution, Thermo Scientific Chemicals
Glycerol, for analysis, 86-88% wt% aqueous solution, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glycerol, for analysis, 86-88% wt% aqueous solution, Thermo Scientific Chemicals
CAS: 56-81-5 | C3H8O3 | 92.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
10 L
Packaging:
View all
Glass bottle
Catalog number FSA295602500
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS762-42-5
IUPAC Name1,4-dimethyl but-2-ynedioate
Molecular FormulaC6H6O4
InChI KeyVHILMKFSCRWWIJ-UHFFFAOYSA-N
SMILESCOC(=O)C#CC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.4460 to 1.4500 (20°C, 589 nm)
GC>=97.5 %