Fullerene C60, 99.9%, Thermo Scientific Chemicals
Fullerene C60, 99.9%, Thermo Scientific Chemicals
Fullerene C60, 99.9%, Thermo Scientific Chemicals
Fullerene C60, 99.9%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Fullerene C60, 99.9%, Thermo Scientific Chemicals
CAS: 99685-96-8 | C60 | 720.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Packaging:
Glass bottle
Catalog number FSA295010010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13209-15-9
IUPAC Name1,2-bis(dibromomethyl)benzene
Molecular FormulaC8H6Br4
InChI KeyLNAOKZKISWEZNY-UHFFFAOYSA-N
SMILESBrC(Br)C1=CC=CC=C1C(Br)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown to pale gray
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Melting Point (clear melt)111.0-120.0?C