Cyclen, 97%, Thermo Scientific Chemicals
Cyclen, 97%, Thermo Scientific Chemicals
Cyclen, 97%, Thermo Scientific Chemicals
Cyclen, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclen, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
50 mg
1 g
Packaging:
Glass bottle
Catalog number FSA294322500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7790-21-8
IUPAC Namepotassium periodate
Molecular FormulaIKO4
InChI KeyFJVZDOGVDJCCCR-UHFFFAOYSA-M
SMILES[K+].[O-][I](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Titration with Na2S2O399.8 to 100.3 % (dried basis)
Other halogens (as Cl)=<0.01 %
Manganese (Mn)=<1 ppm
Chelation/Complexation reagent
RUO – Research Use Only