2,6-Di-tert-butyl-4-methoxyphenol, 97%, Thermo Scientific Chemicals
2,6-Di-tert-butyl-4-methoxyphenol, 97%, Thermo Scientific Chemicals
2,6-Di-tert-butyl-4-methoxyphenol, 97%, Thermo Scientific Chemicals
2,6-Di-tert-butyl-4-methoxyphenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,6-Di-tert-butyl-4-methoxyphenol, 97%, Thermo Scientific Chemicals
CAS: 489-01-0 | C15H24O2 | 236.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA294160250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS563-63-3
IUPAC Namesilver(1+) acetate
Molecular FormulaC2H3AgO2
InChI KeyCQLFBEKRDQMJLZ-UHFFFAOYSA-M
SMILES[Ag+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormMicrocrystalline powder or powder
Assay from Supplier's CofA≥98.5% to ≤101.5% [UK sourced material]
Assay from Supplier's CofA64.5 ± 0.5% Ag w/w [US sourced material]
CommentMaterial sourced either in UK or USA
CommentMay contain trace amount of fine black particles from the manufacturing process