Octyl 4-methoxycinnamate, 98%, stabilized, Thermo Scientific Chemicals
Octyl 4-methoxycinnamate, 98%, stabilized, Thermo Scientific Chemicals
Octyl 4-methoxycinnamate, 98%, stabilized, Thermo Scientific Chemicals
Octyl 4-methoxycinnamate, 98%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Octyl 4-methoxycinnamate, 98%, stabilized, Thermo Scientific Chemicals
CAS: 5466-77-3 | C18H26O3 | 290.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA291160250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS960-16-7
IUPAC Name(2,3,4-tributylphenyl)stannanylidyne
Molecular FormulaC18H29Sn
InChI KeyZMUNITMSSGNSOT-UHFFFAOYSA-N
SMILESCCCCC1=CC=C([Sn])C(CCCC)=C1CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Assay (GC)≥96.0%
Proton NMRConforms