2-Bromoacetamide, 98%, Thermo Scientific Chemicals
2-Bromoacetamide, 98%, Thermo Scientific Chemicals
2-Bromoacetamide, 98%, Thermo Scientific Chemicals
2-Bromoacetamide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromoacetamide, 98%, Thermo Scientific Chemicals
CAS: 683-57-8 | C2H4BrNO | 137.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA291100250
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS130-16-5
IUPAC Name5-chloroquinolin-8-ol
Molecular FormulaC9H6ClNO
InChI KeyCTQMJYWDVABFRZ-UHFFFAOYSA-N
SMILESOC1=C2N=CC=CC2=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥94.0%
Appearance (Color)White to cream to pale green to pale brown to grey
Melting Point (clear melt)120.5-129.5?C