Selenophene, 99%, Thermo Scientific Chemicals
Selenophene, 99%, Thermo Scientific Chemicals
Selenophene, 99%, Thermo Scientific Chemicals
Selenophene, 99%, Thermo Scientific Chemicals
Selenophene, 99%, Thermo Scientific Chemicals
CAS: 288-05-1 | C4H4Se | 131.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA271900100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10284-63-6
IUPAC Name(1R,2S,4S,5S)-6-methoxycyclohexane-1,2,3,4,5-pentol
Molecular FormulaC7H14O6
InChI KeyDSCFFEYYQKSRSV-FEPQRWDDSA-N
SMILESCOC1[C@@H](O)[C@@H](O)C(O)[C@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%