1,4-Dioxane, 99.5%, for analysis, stabilized, Thermo Scientific Chemicals
1,4-Dioxane, 99.5%, for analysis, stabilized, Thermo Scientific Chemicals
1,4-Dioxane, 99.5%, for analysis, stabilized, Thermo Scientific Chemicals
1,4-Dioxane, 99.5%, for analysis, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Dioxane, 99.5%, for analysis, stabilized, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number FSA270490010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS109-49-9
IUPAC Namehex-5-en-2-one
Molecular FormulaC6H10O
InChI KeyRNDVGJZUHCKENF-UHFFFAOYSA-N
SMILESCC(=O)CCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4190-1.4220 @ 20?C
Appearance (Color)Clear colorless to pale yellow